Tisk…

5-Fluorocytosine 99+%

Dodavatel: Thermo Fisher Scientific

Synonyms: 5-FC, 4-Amino-5-fluoro-2-pyrimidinol, Flucytosine, 6-Amino-5-fluoropyrimidin-2(1H)-one

258340010 258340050 258342500
ACRO258340010EA 6220 CZK
ACRO258340010 ACRO258340050 ACRO258342500
5-Fluorocytosine 99+%
5-Fluorocytosine
Vzorec: C₄H₄FN₃O
MW: 129.09
Bod varu: 298
Bod tání: 296 °C (rozkl.)
Storage Temperature: Ambient
MDL Number: MFCD00006035
CAS číslo: 2022-85-7
EINECS: 217-968-7
Merck Index: 13,04151

Objednat

Výsledky specifikačního testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1.0 % (1 g, 105°C)
Titration with HClO4 ≥99.0 % (On dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR