5-Fluorocytosine, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
5-FC, 4-Amino-5-fluoro-2-pyrimidinol, Flucytosine, 6-Amino-5-fluoropyrimidin-2(1H)-one
M-Clarity™ quality level = MQ200
F7129-1G
F7129-5G
SIALF7129-1GEA
11560
CZK
SIALF7129-1G
SIALF7129-5G
5-Fluorocytosine, Sigma-Aldrich®
5-Fluorocytosine
Vzorec:
C₄H₄FN₃O MW: 129,09 g/mol Bod varu: 298 °C (1013 hPa) Bod tání: 165 °C |
MDL Number:
MFCD00006035 CAS číslo: 2022-85-7 EINECS: 217-968-7 Merck Index: 13,04151 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...