Tisk…

4-(Bromomethyl)phenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

679437-1G 679437-5G
SIAL679437-1GEA 3350 CZK
SIAL679437-1G SIAL679437-5G
4-(Bromomethyl)phenylboronic acid, Sigma-Aldrich®
4-(Bromomethyl)phenylboronic acid
Vzorec: C₇H₈BBrO₂
MDL Number: MFCD01318113
CAS číslo: 68162-47-0
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR