3-Tolylboronic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(3-Methylphenyl)metaboric acid, Dihydroxy(3-methylphenyl)borane, 3-Methylbenzeneboronic acid, 3-Methylphenylboronic acid, m-Tolylboronic acid
M-Clarity™ quality level = MQ100
393614-1G
393614-5G
SIAL393614-1GEA
1170
CZK
SIAL393614-1G
SIAL393614-5G
3-Tolylboronic acid, Sigma-Aldrich®
3-Tolylboronic acid
Vzorec:
C₇H₉BO₂ MW: 135,96 g/mol Bod tání: 165 °C |
MDL Number:
MFCD00040198 CAS číslo: 17933-03-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...