Tisk…

3-Aminophenylboronic acid monohydrate 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Dihydroxy(3-aminophenyl)borane monohydrate, 3-Aminobenzeneboronic acid monohydrate, 3-Aminobenzeneboronic acid, 3-Aminophenylboronic Acid Monohydrate, (3-Aminophenyl)metaboric acid monohydrate

A18189.03 A18189.06
A18189.03EA 1860 CZK
A18189.03 A18189.06
3-Aminophenylboronic acid monohydrate 97%
3-Aminophenylboronic acid monohydrate

Purity: 97% CAS No.: 206658-89-1 Molecular Formula: C6H8BNO2.H2O Formula Weight: 154.96 (136.95anhy) Melting Point: 93-97° Beilstein Reference: 2936342

Vzorec: C₆H₈BNO₂·1H₂O
Bod tání: 93…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00149554
CAS číslo: 206658-89-1
EINECS: 250-189-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR