Tisk…

Citral cis- and trans-mixture 95%

Dodavatel: Thermo Fisher Scientific
Varování

A16169.18 A16169.22 A16169.36
A16169.18EA 872 CZK
A16169.18 A16169.22 A16169.36
Citral cis- and trans-mixture 95%
Citral

Purity: 95% CAS No.: 5392-40-5 Molecular Formula: C10H16O Formula Weight: 152.24 Storage Temperature: 2°C to 8°C Flash Point: 98°(208°F) Refractive Index: 1.4880 Melting Point: -10° Boiling Point: 228-229° Beilstein Reference: 1721871 Merck Reference: 14,2322 MDL No.: MFCD00006997

Vzorec: C₁₀H₁₆O
Bod varu: 228…229 °C
Bod tání: < –10 °C
Hustota: 0,888
Bod vzplanutí: 98 °C (208 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00006997
CAS číslo: 5392-40-5
EINECS: 226-394-6
Merck Index: 14,2322

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR