3,5-Bis(trifluoromethyl)phenylboronic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3,5-Bis(trifluoromethyl)benzeneboronic acid, (3,5-Bis(trifluoromethyl)phenyl)metaboric acid, (3,5-Bis(trifluoromethyl)phenyl)boronic acid, Dihydroxy(3,5-bis(trifluoromethyl)phenyl)borane
A11373.03
A11373.06
A11373.14
A11373.03EA
1130
CZK
A11373.03
A11373.06
A11373.14
3,5-Bis(trifluoromethyl)phenylboronic acid ≥97%
3,5-Bis(trifluoromethyl)phenylboronic acid
Purity: 97+%
CAS No.: 73852-19-4
Molecular Formula: C8H5BF6O2
Formula Weight: 257.93
Melting Point: 219-224°
Beilstein Reference: 7379990
Vzorec:
C₈H₅BF₆O₂ MW: 257,93 g/mol Bod tání: 219…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051850 CAS číslo: 73852-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...