2-Phenoxynicotinic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A11247.03
A11247.06
A11247.14
A11247.03EA
759
CZK
A11247.03
A11247.06
A11247.14
2-Phenoxynicotinic acid ≥98%
2-Phenoxynicotinic acid
Purity: 98%
CAS No.: 35620-71-4
Molecular Formula: C12H9NO3
Formula Weight: 215.21
Melting Point: 179-182°
Beilstein Reference: 172906
Vzorec:
C₁₂H₉NO₃ Bod tání: 179…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014629 CAS číslo: 35620-71-4 EINECS: 252-647-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...