Tisk…

2-Methylnicotinic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Methylpyridine-3-carboxylic acid, 2-Methyl-3-pyridinecarboxylic acid

M-Clarity™ quality level = MQ100

325228-1G 325228-5G
SIAL325228-1GEA 2190 CZK
SIAL325228-1G SIAL325228-5G
2-Methylnicotinic acid, Sigma-Aldrich®
2-Methylnicotinic acid
Vzorec: C₇H₇NO₂
MW: 137,14 g/mol
MDL Number: MFCD00013449
CAS číslo: 3222-56-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR