Tisk…

2-Methoxynicotinic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Methoxy-3-pyridinecarboxylic acid, 2-Methoxypyridine-3-carboxylic acid

M-Clarity™ quality level = MQ100

632767-1G 632767-5G
SIAL632767-1GEA 607 CZK
SIAL632767-1G SIAL632767-5G
2-Methoxynicotinic acid, Sigma-Aldrich®
2-Methoxynicotinic acid
Vzorec: C₇H₇NO₃
MW: 153,14 g/mol
MDL Number: MFCD00085110
CAS číslo: 16498-81-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR