Tisk…

Iron(III) citrate monohydrate 18-20% (Fe basis), Supelco®

Dodavatel: Merck

Synonyms: Iron (III) citrate tribasic monohydrate, Ferric citrate monohydrate, Ferric Citrate, n-Hydrate

M-Clarity™ quality level = MQ100

44941-1KG 44941-2.5KG 44941-250G 44941
44941-1KGEA 4940 CZK
44941-1KG 44941-2.5KG 44941-250G LACH44941
Iron(III) citrate monohydrate 18-20% (Fe basis), Supelco®
Iron(III) citrate monohydrate
Vzorec: C₆H₅FeO₇·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00013098
CAS číslo: 2338-05-8
EINECS: 249-117-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c3ffd2085fc47373ffb977a8f2aa6a56bb0f95d&vid=9e2b17117cfa224eab3aa5eb515187755f2495fe&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR