Tisk…

2-Aminophenylboronic acid 96%

Dodavatel: Thermo Fisher Scientific
Varování

L18069.03 L18069.06 L18069.MC
L18069.03EA 6250 CZK
L18069.03 L18069.06 L18069.MC
2-Aminophenylboronic acid 96%
2-Aminophenylboronic acid

Purity: 96% CAS No.: 5570-18-3 Molecular Formula: C6H8BNO2 Formula Weight: 136.95 Melting Point: 188-190° Beilstein Reference: 2962939

Vzorec: C₆H₈BNO₂
Bod tání: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD01074645
CAS číslo: 5570-18-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR