5-Acetyl-2,4-dimethylthiazole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,4-Dimethyl-5-acetylthiazole, 1-(2,4-Dimethylthiazol-5-yl)ethanone, 1-(2,4-Dimethylthiazol-5-yl)ethan-1-one
M-Clarity™ quality level = MQ300
W326704-100G-K
W326704-1KG-K
W326704-25G-K
SIALW326704-100G-KEA
0
CZK
SIALW326704-100G-K
SIALW326704-1KG-K
SIALW326704-25G-K
5-Acetyl-2,4-dimethylthiazole, Sigma-Aldrich®
5-Acetyl-2,4-dimethylthiazole
Vzorec:
C₇H₉NOS MW: 155,22 g/mol Bod varu: 228 °C (1013 hPa) Hustota: 1,15 g/cm³ (20 °C) |
MDL Number:
MFCD00005331 CAS číslo: 38205-60-6 EINECS: 253-826-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...