2,4-Bis(trifluoromethyl)phenylboronic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
L18161.06
L18161.03
L18161.06EA
13940
CZK
L18161.06
L18161.03
2,4-Bis(trifluoromethyl)phenylboronic acid ≥97%
2,4-Bis(trifluoromethyl)phenylboronic acid
Purity: 97%
CAS No.: 153254-09-2
Molecular Formula: C8H5BF6O2
Formula Weight: 257.93
MDL No.: MFCD01631349
Vzorec:
C₈H₅BF₆O₂ MW: 257,93 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631349 CAS číslo: 153254-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...