Tisk…

Tropone, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

252832-1G 252832-5G
SIAL252832-1GEA 2090 CZK
SIAL252832-1G SIAL252832-5G
Tropone, Sigma-Aldrich®
Tropone
Vzorec: C₇H₆O
MDL Number: MFCD00014331
CAS číslo: 539-80-0
EINECS: 208-725-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR