Tropone ≥97%
Dodavatel: Thermo Fisher Scientific
L00473.03
L00473.06
L00473.03EA
1830
CZK
L00473.03
L00473.06
Tropone ≥97%
Tropone
Purity: 97%
CAS No.: 539-80-0
Molecular Formula: C7H6O
Formula Weight: 106.12
Storage Temperature: -30°C to -10°C
Flash Point:
Vzorec:
C₇H₆O Bod varu: 91…92 °C (4 mmHg) Hustota: 1,094 Bod vzplanutí: >110 °C (230 °F) |
MDL Number:
MFCD00014331 CAS číslo: 539-80-0 EINECS: 208-725-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...