2,2'-Thiodiacetic acid 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Thiodiglycolic acid, Thiodiacetic Acid
138740010
138740250
138741000
ACRO138740010EA
6000
CZK
ACRO138740010
ACRO138740250
ACRO138741000
2,2'-Thiodiacetic acid 98%
2,2'-Thiodiacetic acid
Vzorec:
C₄H₆O₄S MW: 150,16 g/mol Bod varu: 168 °C (14 mmHg) Bod tání: 128…131 °C Hustota: 1,661 g/cm³ (20 °C) |
MDL Number:
MFCD00004359 CAS číslo: 123-93-3 EINECS: 204-663-9 UN: 3261 ADR: 8,III Merck Index: 12,09467 |
Výsledky specifikačního testu
Appearance | White to grey Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 128 to 131 °C |
Titration with NaOH | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Specific optical rotation | -36.6° to -40.6° (20 °C, 589 nm) (c=1,4 in DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...