Tisk…

2,2'-Thiodiacetic acid ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Thiodiglycolic acid, Thiodiacetic Acid

A17880.14 A17880.22 A17880.36
A17880.14EA 824 CZK
A17880.14 A17880.22 A17880.36
2,2'-Thiodiacetic acid ≥98%
2,2'-Thiodiacetic acid

Purity: 98% CAS No.: 123-93-3 Molecular Formula: C4H6O4S Formula Weight: 150.15 Melting Point: 127-131° Beilstein Reference: 1764392 Merck Reference: 14,9331 UN No.: 3261 MDL No.: MFCD00004359

Vzorec: C₄H₆O₄S
MW: 150,16 g/mol
Bod varu: 168 °C (14 mmHg)
Bod tání: 127…131 °C
Hustota: 1,661 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004359
CAS číslo: 123-93-3
EINECS: 204-663-9
UN: 3261
ADR: 8,III
Merck Index: 14,9331

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR