2-Nonyn-1-ol 96%
Dodavatel: Thermo Fisher Scientific
L06380.06
L06380.14
L06380.06EA
3300
CZK
L06380.06
L06380.14
2-Nonyn-1-ol 96%
2-Nonyn-1-ol
Purity: 96%
CAS No.: 5921-73-3
Molecular Formula: CH3(CH2)5C≡CCH2OH
Formula Weight: 140.23
Refractive Index: 1.4560
Boiling Point: 109-111°/11mm
MDL No.: MFCD00041566
Vzorec:
C₉H₁₆O Bod varu: 109…111 °C (11 mmHg) Hustota: 0,873 Storage Temperature: Ambient |
MDL Number:
MFCD00041566 CAS číslo: 5921-73-3 EINECS: 227-646-8 |