4-Fluoro-2-methylbenzyl alcohol ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(4-Fluoro-2-methylphenyl)methanol
H32364.03
H32364.06
H32364.03EA
2010
CZK
H32364.03
H32364.06
4-Fluoro-2-methylbenzyl alcohol ≥99%
4-Fluoro-2-methylbenzyl alcohol
Purity: 99%
CAS No.: 80141-91-9
Molecular Formula: C8H9FO
Formula Weight: 140.16
MDL No.: MFCD03701058
Vzorec:
C₈H₉FO MW: 140.16 Storage Temperature: Ambient |
MDL Number:
MFCD03701058 CAS číslo: 80141-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...