Tisk…

2-Fluoro-4-methoxybenzyl alcohol ≥97%

Dodavatel: Thermo Fisher Scientific
Varování

H32118.03 H32118.09
H32118.03EA 1800 CZK
H32118.03 H32118.09
2-Fluoro-4-methoxybenzyl alcohol ≥97%
2-Fluoro-4-methoxybenzyl alcohol

Purity: 97% CAS No.: 405-09-4 Molecular Formula: C8H9FO2 Formula Weight: 156.15 MDL No.: MFCD04116336

Vzorec: C₈H₉FO₂
MW: 156.16
Storage Temperature: Ambient
MDL Number: MFCD04116336
CAS číslo: 405-09-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR