Diethylenglykolmonoethylether, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-(2-Ethoxyethoxy)ethanol, Di(ethylene glycol) ethyl ether, Diethylene Glycol Monoethyl Ether
M-Clarity™ quality level = MQ300
W509043-10KG
W509043-1KG
W509043-25KG
W509043-SAMPLE
SIALW509043-10KGEA
14940
CZK
SIALW509043-10KG
SIALW509043-1KG
SIALW509043-25KG
SIALW509043-SAMPLE
Diethylenglykolmonoethylether, Sigma-Aldrich®
Diethylenglykolmonoethylether
Vzorec:
C₆H₁₄O₃ MW: 134,18 g/mol Bod varu: 197 °C (1013 hPa) Bod tání: –80 °C Hustota: 0,99 g/cm³ (20 °C) Bod vzplanutí: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002872 CAS číslo: 111-90-0 EINECS: 203-919-7 REACH: 01-2119475105-42 Merck Index: 13,01809 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...