4-Chloro-3,5-xylenol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Chloro-5-hydroxy-3-xylene, Chloroxylenol, 4-Chloro-3,5-dimethylphenol
B21964.22
B21964.36
B21964.22EA
755
CZK
B21964.22
B21964.36
4-Chloro-3,5-xylenol ≥98%
4-Chloro-3,5-xylenol
Purity: 98+%
CAS No.: 88-04-0
Molecular Formula: C8H9ClO
Formula Weight: 156.61
Flash Point: 138°(280°F)
Odour: Odourless
Boiling Point: ca 246°
Beilstein Reference: 1862539
Merck Reference: 14,2176
MDL No.: MFCD00002324
Vzorec:
C₈H₉ClO MW: 156.61 Bod varu: ∼246 °C Bod tání: 112…116 °C Hustota: 1.364 g/cm³ (20 °C) Bod vzplanutí: 138 °C (280 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002324 CAS číslo: 88-04-0 EINECS: 201-793-8 Merck Index: 13,02194 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...