Tisk…

4-Bromo-2-methoxybenzyl alcohol, Sigma-Aldrich®

Dodavatel: Merck

653187-10G 653187-1G
SIAL653187-10GEA 2220 CZK
SIAL653187-10G SIAL653187-1G
4-Bromo-2-methoxybenzyl alcohol, Sigma-Aldrich®
4-Bromo-2-methoxybenzyl alcohol
Vzorec: C₈H₉BrO₂
MW: 217,06 g/mol
MDL Number: MFCD07369762
CAS číslo: 17102-63-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR