4-Bromo-2,6-xylenol ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Bromo-2,6-dimethylphenol, 5-Bromo-2-hydroxy-3-xylene
B21669.14
B21669.22
B21669.36
B21669.14EA
1080
CZK
B21669.14
B21669.22
B21669.36
4-Bromo-2,6-xylenol ≥99%
4-Bromo-2,6-xylenol
Purity: 99%
CAS No.: 2374-05-2
Molecular Formula: C8H9BrO
Formula Weight: 201.07
Odour: Phenol-like
Melting Point: 74-78°
Beilstein Reference: 1862399
MDL No.: MFCD00002314
Vzorec:
C₈H₉BrO MW: 201,06 g/mol Bod varu: 259 °C (1013 hPa) Bod tání: 74…78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002314 CAS číslo: 2374-05-2 EINECS: 219-153-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...