4-Amino-m-cresol 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
B23152.14
B23152.22
AB131401.0025
B23152.14EA
1510
CZK
B23152.14
B23152.22
ABCRAB131401.0025
4-Amino-m-cresol 98%
4-Amino-m-cresol
Purity: 98%
CAS No.: 2835-99-6
Molecular Formula: C7H9NO
Formula Weight: 123.16
Melting Point: 176-179°
Beilstein Reference: 2078803
UN No.: 3077
MDL No.: MFCD00007871
Vzorec:
C₇H₉NO Bod varu: 259 °C (1013 hPa) Bod tání: 176…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007871 CAS číslo: 2835-99-6 EINECS: 220-621-2 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...