Tisk…

2-Amino-m-cresol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Hydroxy-6-methylaniline, 2-Amino-3-hydroxytoluene, 2-Amino-3-methylphenol, 1-Amino-2-hydroxy-6-methylbenzene

M-Clarity™ quality level = MQ100

336572-25G 336572-5G
SIAL336572-25GEA 4900 CZK
SIAL336572-25G SIAL336572-5G
2-Amino-m-cresol, Sigma-Aldrich®
2-Amino-m-cresol
Vzorec: C₇H₉NO
MW: 123,15 g/mol
MDL Number: MFCD00075082
CAS číslo: 2835-97-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR