Tisk…

2-Amino-p-cresol 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 3-Amino-4-hydroxytoluene, 1-Amino-2-hydroxy-5-methylbenzene, 2-Amino-4-methylphenol, 2-Hydroxy-5-methylaniline

A19546.18 A19546.30
A19546.18EA 968 CZK
A19546.18 A19546.30
2-Amino-p-cresol 98%
2-Amino-p-cresol

Purity: 98% CAS No.: 95-84-1 Molecular Formula: C7H9NO Formula Weight: 123.16 Melting Point: 134-138° Beilstein Reference: 606494

Vzorec: C₇H₉NO
MW: 123,15 g/mol
Bod varu: 259 °C (1013 hPa)
Bod tání: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00007699
CAS číslo: 95-84-1
EINECS: 202-457-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR