Tisk…

6-Amino-m-cresol 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 1-Amino-2-hydroxy-4-methylbenzene, 2-Hydroxy-4-methylaniline, Cetilistat Impurity 4, 4-Amino-3-hydroxytoluene, 2-Amino-5-methylphenol

B23560.09 B23560.18
B23560.09EA 944 CZK
B23560.09 B23560.18
6-Amino-m-cresol 98%
6-Amino-m-cresol

Purity: 98% CAS No.: 2835-98-5 Molecular Formula: C7H9NO Formula Weight: 123.16 Melting Point: 155-159° Beilstein Reference: 386144

Vzorec: C₇H₉NO
MW: 123,15 g/mol
Bod varu: 259 °C (1013 hPa)
Bod tání: 155…159 °C
Storage Temperature: Ambient
MDL Number: MFCD00007693
CAS číslo: 2835-98-5
EINECS: 220-620-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR