Tisk…

4-Iodo-2,5-xylenol ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Iodo-2,5-dimethylphenol, 2-Hydroxy-5-iodo-4-xylene, 5-Hydroxy-2-iodo-4-xylene, 2,5-Dimethyl-4-iodophenol

H50565.03 H50565.06
H50565.03EA 2080 CZK
H50565.03 H50565.06
4-Iodo-2,5-xylenol ≥98%
4-Iodo-2,5-xylenol

Purity: 98% CAS No.: 114971-53-8 Molecular Formula: C8H9IO Formula Weight: 248.06 MDL No.: MFCD00219922

Vzorec: C₈H₉IO
MW: 248,06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00219922
CAS číslo: 114971-53-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR