Adiponitrile 99%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
113830010
113831000
ACRO113830010EA
3030
CZK
ACRO113830010
ACRO113831000
Adiponitrile 99%
Adiponitrile
Vzorec:
C₆H₈N₂ Bod varu: 296 °C (1013 hPa) Bod tání: 1…3 °C Hustota: 0,95 g/cm³ Bod vzplanutí: 163 °C |
MDL Number:
MFCD00001975 CAS číslo: 111-69-3 EINECS: 203-896-3 UN: 2205 ADR: 6.1,III |
Výsledky specifikačního testu
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4370 to 1.4390 (20°C, 589 nm) |
GC | ≥98.5 % |
Microbial contamination | (Salmonella) Absent in 25 g |
pH | 6 to 8 (50 % suspension in water) |
Sulfite (as SO2) | ≤10 ppm |
Arsenic (As) | ≤0.07 ppm |
Cadmium (Cd) | ≤0.05 ppm |
Iron (Fe) | ≤10 ppm |
Lead (Pb) | ≤0.1 ppm |
Mercury (Hg) | ≤0.05 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...