Tisk…

Adiponitrile ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Hexanedinitrile

Packaged under argon in resealable ChemSeal™ bottles.

43679.AE 43679.AP
43679.AEEA 1040 CZK
43679.AE 43679.AP
Adiponitrile ≥99%
Adiponitrile

Packaged under Argon in resealable ChemSeal® bottles


Purity: 99+% CAS No.: 111-69-3 Molecular Formula: NC(CH2)4CN Formula Weight: 108.14 Flash Point: 163°(325°F) Physical Form: Liquid Density: 0.951 Refractive Index: 1.4380 Odour: Odourless Melting Point: 2° Boiling Point: 295° Beilstein Reference: 1740005 UN No.: 2205 MDL No.: MFCD00001975

Vzorec: C₆H₈N₂
MW: 108.14
Bod varu: 295 °C
Bod tání: 2
Hustota: 0.951
Bod vzplanutí: 163 °C (325 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001975
CAS číslo: 111-69-3
EINECS: 203-896-3
UN: 2205
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR