2,6-Dimethoxy-p-cresol, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3,5-Dimethoxy-4-hydroxytoluene, 4-Methyl-2,6-dimethoxyphenol, 1,3-Dimethoxy-2-hydroxy-5-methylbenzene, 2,6-Dimethoxy-4-methylphenol
M-Clarity™ quality level = MQ300
W370401-1KG
W370401-25G
W370401-SAMPLE
SIALW370401-1KGEA
0
CZK
SIALW370401-1KG
SIALW370401-25G
SIALW370401-SAMPLE
2,6-Dimethoxy-p-cresol, Sigma-Aldrich®
2,6-Dimethoxy-p-cresol
Vzorec:
C₉H₁₂O₃ MW: 168,19 g/mol |
MDL Number:
MFCD00017289 CAS číslo: 6638-05-7 EINECS: 229-641-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...