3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
446424-25G
446424-5G
SIAL446424-25GEA
7760
CZK
SIAL446424-25G
SIAL446424-5G
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, Sigma-Aldrich®
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol
Vzorec:
C₁₅H₂₄O₂ MW: 236,35 g/mol |
MDL Number:
MFCD00017254 CAS číslo: 88-26-6 EINECS: 201-815-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...