Tisk…

(±)-1,2-Octanediol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane

M-Clarity™ quality level = MQ200

213705-10G 213705-50G
SIAL213705-10GEA 832 CZK
SIAL213705-10G SIAL213705-50G
(±)-1,2-Octanediol, Sigma-Aldrich®
(±)-1,2-Octanediol
Vzorec: C₈H₁₈O₂
MW: 146,23 g/mol
Bod varu: 243 °C (1013 hPa)
Bod tání: 36 °C
MDL Number: MFCD00010738
CAS číslo: 1117-86-8
EINECS: 214-254-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR