Tisk…

Triethylenglykol ≥99%

Dodavatel: Thermo Scientific

Synonyms: 2,2'-(Ethylenedioxy)diethanol, 2,2'-(Ethane-1,2-diylbis(oxy))diethanol

A10772.0D A10772.0F A10772.AP
A10772.0DEA 7620 CZK
A10772.0D A10772.0F A10772.AP
Triethylenglykol ≥99%
Triethylenglykol

Purity: 99% CAS No.: 112-27-6 Molecular Formula: C6H14O4 Formula Weight: 150.17 Flash Point: 165°(329°F) Physical Form: Liquid Density: 1.125 Refractive Index: 1.4555 Sensitivity: Hygroscopic Appearance: Colourless Odour: Nearly Odourless Melting Point: -7° Boiling Point: 284-288° Beilstein Reference: 969357 Merck Reference: 14,9670 MDL No.: MFCD00002880

Vzorec: C₆H₁₄O₄
MW: 150,17 g/mol
Bod varu: 284…288 °C
Bod tání: –7 °C
Hustota: 1,125
Bod vzplanutí: 165 °C (329 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002880
CAS číslo: 112-27-6
EINECS: 203-953-2
Merck Index: 14,9670

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR