Tisk…

Citral dimethyl acetal, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W230502-100G-K W230502-1KG-K W230502-4KG-K
SIALW230502-100G-KEA 2600 CZK
SIALW230502-100G-K SIALW230502-1KG-K SIALW230502-4KG-K
Citral dimethyl acetal, Sigma-Aldrich®
Citral dimethyl acetal
Vzorec: C₁₂H₂₂O₂
MDL Number: MFCD00008909
CAS číslo: 7549-37-3
EINECS: 231-434-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR