Tisk…

tert-Butanol-[D10], Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Trimethylcarbinol-D10, tert-Butyl alcohol-D10, 2-Methyl-2-propanol-D10

M-Clarity™ quality level = MQ200

175889-1G 175889-5G
SIAL175889-1GEA 5490 CZK
SIAL175889-1G SIAL175889-5G
tert-Butanol-[D10], Sigma-Aldrich®
tert-Butanol-[D10]
Vzorec: D₃CC(CD₃)(OD)CD₃
MW: 84.04
Bod varu: 82
Storage Temperature: Ambient
CAS číslo: 53001-22-2
EINECS: 258-288-0
UN: 1120
ADR: 3 II

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR