D(-)-Ribóza, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W379301-100G
W379301-1KG
W379301-5KG
W379301-SAMPLE
SIALW379301-100GEA
4280
CZK
SIALW379301-100G
SIALW379301-1KG
SIALW379301-5KG
SIALW379301-SAMPLE
D(-)-Ribóza, Sigma-Aldrich®
D(-)-Ribose
Vzorec:
C₅H₁₀O₅ MW: 150,13 g/mol Bod varu: 331 °C (1013 hPa) Bod tání: 90…95 °C Hustota: 0,8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00135453 CAS číslo: 50-69-1 EINECS: 200-059-4 Merck Index: 12,08371 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...