4-Methylthiazole, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W371602-100G-K
W371602-1KG-K
W371602-5KG-K
SIALW371602-100G-KEA
6780
CZK
SIALW371602-100G-K
SIALW371602-1KG-K
SIALW371602-5KG-K
4-Methylthiazole, Sigma-Aldrich®
4-Methylthiazole
Vzorec:
C₄H₅NS MW: 99,16 g/mol Bod varu: 133 °C (1013 hPa) Hustota: 1,1187 g/cm³ (20 °C) |
MDL Number:
MFCD00005340 CAS číslo: 693-95-8 EINECS: 211-764-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...