Tisk…

2-Methylbutyl isovalerate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Isopropyl acetic acid 2-methylbutyl ester, Isovaleric acid 2-methylbutyl ester, 2-Methylbutyl isopentanoate, 3-Methylbutyric acid 2-methylbutyl ester, 3-Methylbutanoic acid 2-methylbutyl ester

M-Clarity™ quality level = MQ300

W350605-1KG-K W350605-4KG-K W350605-9KG-K
SIALW350605-1KG-KEA 4160 CZK
SIALW350605-1KG-K SIALW350605-4KG-K SIALW350605-9KG-K
2-Methylbutyl isovalerate, Sigma-Aldrich®
2-Methylbutyl isovalerate
Vzorec: C₁₀H₂₀O₂
MW: 172.27
CAS číslo: 2445-77-4
EINECS: 219-496-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR