Ethyl crotonate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
trans-Crotonic acid ethyl ester, 2-Butenoic acid ethyl ester, Crotonic acid ethyl ester, Ethyl trans-2-butenoate, trans-Ethyl crotonate, trans-2-Butenoic acid ethyl ester
M-Clarity™ quality level = MQ300
W348600-10KG-K
W348600-1KG-K
W348600-20KG-K
SIALW348600-10KG-KEA
40920
CZK
SIALW348600-10KG-K
SIALW348600-1KG-K
SIALW348600-20KG-K
Ethyl crotonate, Sigma-Aldrich®
Ethyl crotonate
Vzorec:
C₆H₁₀O₂ MW: 114.14 Bod varu: 143 Bod tání: –79 °C Hustota: 0.9208 g/cm³ (20 °C) Bod vzplanutí: 2 |
MDL Number:
MFCD00009289 CAS číslo: 623-70-1 EINECS: 210-808-7 UN: 1862 ADR: 3 II Merck Index: 13,02625 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...