Tisk…

4,5-Dimethylthiazole, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W327409-100G W327409-1KG W327409-25G W327409-SAMPLE
SIALW327409-100GEA 7950 CZK
SIALW327409-100G SIALW327409-1KG SIALW327409-25G SIALW327409-SAMPLE
4,5-Dimethylthiazole, Sigma-Aldrich®
4,5-Dimethylthiazole
Vzorec: C₅H₇NS
MW: 113,18 g/mol
Bod varu: 149…153 °C (1013 hPa)
Hustota: 1,0828 g/cm³ (20 °C)
MDL Number: MFCD00005336
CAS číslo: 3581-91-7
EINECS: 222-703-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR