Benzothiazole, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Benzo[d]thiazole
M-Clarity™ quality level = MQ300
W325600-10KG
W325600-1KG
W325600-25KG
W325600-SAMPLE
SIALW325600-10KGEA
47570
CZK
SIALW325600-10KG
SIALW325600-1KG
SIALW325600-25KG
SIALW325600-SAMPLE
Benzothiazole, Sigma-Aldrich®
Benzothiazole
Vzorec:
C₇H₅NS MW: 135,19 g/mol Bod varu: 231 °C (1013 hPa) Hustota: 1,246 g/cm³ (20 °C) |
MDL Number:
MFCD00005775 CAS číslo: 95-16-9 EINECS: 202-396-2 UN: 2810 ADR: 6.1,III Merck Index: 13,01108 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...