2-(4-Methylthiazol-5-yl)ethanol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ300
W320404-10KG-K
W320404-1KG-K
W320404-250G-K
W320404-5KG-K
SIALW320404-10KG-KEA
45940
CZK
SIALW320404-10KG-K
SIALW320404-1KG-K
SIALW320404-250G-K
SIALW320404-5KG-K
2-(4-Methylthiazol-5-yl)ethanol, Sigma-Aldrich®
2-(4-Methylthiazol-5-yl)ethanol
Vzorec:
C₆H₉NOS Bod varu: 135 °C (1013 hPa) Hustota: 1,199 g/cm³ (20 °C) |
MDL Number:
MFCD00005339 CAS číslo: 137-00-8 EINECS: 205-272-6 Merck Index: 12,06208 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...