trans-2-Methyl-2-pentenoic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(E)-2-Methylpent-2-enoic acid
M-Clarity™ quality level = MQ300
W319503-100G
W319503-10KG
W319503-1KG
W319503-5KG
W319503-SAMPLE
SIALW319503-100GEA
4130
CZK
SIALW319503-100G
SIALW319503-10KG
SIALW319503-1KG
SIALW319503-5KG
SIALW319503-SAMPLE
trans-2-Methyl-2-pentenoic acid, Sigma-Aldrich®
trans-2-Methyl-2-pentenoic acid
Vzorec:
C₆H₁₀O₂ MW: 114,14 g/mol Bod varu: 214 °C (1013 hPa) Bod tání: 25 °C Hustota: 0,987 g/cm³ (20 °C) |
MDL Number:
MFCD00002655 CAS číslo: 16957-70-3 EINECS: 241-026-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...