Tisk…

2-Isobutylthiazole, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W313408-100G-K W313408-1KG-K
SIALW313408-100G-KEA 12400 CZK
SIALW313408-100G-K SIALW313408-1KG-K
2-Isobutylthiazole, Sigma-Aldrich®
2-Isobutylthiazole
Vzorec: C₇H₁₁NS
MW: 141,24 g/mol
Bod varu: 180 °C (1013 hPa)
Hustota: 0,9953 g/cm³ (25 °C)
MDL Number: MFCD00005334
CAS číslo: 18640-74-9
EINECS: 242-470-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR