Tisk…

(±)-γ-Valerolactone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (±)-4-Hydroxypentanoic acid lactone, (±)-Pentan-4-olide, γ-Pentanolactone, 5-Methyldihydrofuran-2(3H)-one, gamma-Valerolactone

M-Clarity™ quality level = MQ300

W310301-10KG-K W310301-1KG-K W310301-5KG-K
SIALW310301-10KG-KEA 27240 CZK
SIALW310301-10KG-K SIALW310301-1KG-K SIALW310301-5KG-K
(±)-γ-Valerolactone, Sigma-Aldrich®
(±)-γ-Valerolactone
Vzorec: C₅H₈O₂
MW: 100,12 g/mol
Bod varu: 207 °C (1013 hPa)
Bod tání: –31 °C
Hustota: 1,05 g/cm³ (20 °C)
MDL Number: MFCD00005400
CAS číslo: 108-29-2
EINECS: 203-569-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR