4-(4-Methoxyphenyl)-2-butanone, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
1-(4-Methoxyphenyl)-3-butanone, Anisylacetone
M-Clarity™ quality level = MQ300
W267201-100G-K
W267201-1KG-K
W267201-5KG-K
SIALW267201-100G-KEA
2180
CZK
SIALW267201-100G-K
SIALW267201-1KG-K
SIALW267201-5KG-K
4-(4-Methoxyphenyl)-2-butanone, Sigma-Aldrich®
4-(4-Methoxyphenyl)-2-butanone
Vzorec:
C₁₁H₁₄O₂ MW: 178,23 g/mol Bod varu: 72 °C (0,02 mmHg) Bod tání: 10 °C Hustota: 1,0619 g/cm³ (0 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008791 CAS číslo: 104-20-1 EINECS: 203-184-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...