Furfuryl mercaptan, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2-Furanmethanethiol, 2-Furanmethanthiol, 2-Furylmethanethiol
M-Clarity™ quality level = MQ300
W249300-1KG-K
W249300-2.5KG-
SIALW249300-1KG-KEA
5630
CZK
SIALW249300-1KG-K
SIALW249300-2.5KG-
Furfuryl mercaptan, Sigma-Aldrich®
Furfuryl mercaptan
Vzorec:
C₅H₆OS MW: 114,17 g/mol Bod varu: 155 °C (1013 hPa) Hustota: 1,1339 g/cm³ (20 °C) Bod vzplanutí: 51 °C |
MDL Number:
MFCD00003254 CAS číslo: 98-02-2 EINECS: 202-628-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...